Learn More
Biotang Inc Streptavidin-NC Recombinant, produced in E.Coli having a molecular mass of 24kDa. Recombinant Streptavidin-NC not only binds to nitrocellulose membrane readily but also preserves the full biotin binding ability.
Catalog No. 50151074 Shop All BIOTANG Inc Products
Encompass_Preferred
Streptavidin-NC Recombinant, produced in E.Coli having a molecular mass of 24kDa. Recombinant Streptavidin-NC not only binds to nitrocellulose membrane readily but also preserves the full biotin binding ability.
Catalog No. 50-151-074 Supplier Biotang Inc Supplier No. BTPRO338
Streptavidin-NC Recombinant, produced in E.Coli having a molecular mass of 24kDa. Recombinant Streptavidin-NC not only binds to nitrocellulose membrane readily but also preserves the full biotin binding ability.