Learn More
Biotang Inc Streptokinase Recombinant,produced in E.Coli is a non-glycosylated polypeptide chain containing 414 amino acids and having a molecular weight of 47 kd.
Catalog No. 50151075 Shop All BIOTANG Inc Products
Encompass_Preferred
Streptokinase Recombinant,produced in E.Coli is a non-glycosylated polypeptide chain containing 414 amino acids and having a molecular weight of 47 kd.
Catalog No. 50-151-075 Supplier Biotang Inc Supplier No. BTENZ315
Streptokinase Recombinant,produced in E.Coli is a non-glycosylated polypeptide chain containing 414 amino acids and having a molecular weight of 47 kd.