Learn More
Apexbio Technology LLC REC SWINE GM-CSF HIS-FLAG-1mg
SDP
Catalog No. 5000897666
Encompass_Preferred
Recombinant Swine GM-CSF (His Flag) is a specialized protein used in biomedical research primarily in the field of immunology and cell biology Originating from swine it functions by stimulating the growth and differentiation of hematopoietic cells especially targeting granulocyte-macrophage pathways Its mechanism involves binding to specific receptors on target cells facilitating a cascade of intracellular signals Commonly its in vitro activity is observed in the nanomolar to low micromolar range effective in various experimental setups It is typically applied in cell culture models and animal studies to investigate immune responses inflammation and potential therapeutic applications through drug screening Experimental concentrations typically depend on the specific research design allowing flexibility in use across different assays and studies
Catalog No. 50-008-97666 Supplier Apexbio Technology LLC Supplier No. P25591
Add to Cart
Add to Cart
Recombinant Swine GM-CSF (His Flag) is a specialized protein used in biomedical research primarily in the field of immunology and cell biology Originating from swine it functions by stimulating the growth and differentiation of hematopoietic cells especially targeting granulocyte-macrophage pathways Its mechanism involves binding to specific receptors on target cells facilitating a cascade of intracellular signals Commonly its in vitro activity is observed in the nanomolar to low micromolar range effective in various experimental setups It is typically applied in cell culture models and animal studies to investigate immune responses inflammation and potential therapeutic applications through drug screening Experimental concentrations typically depend on the specific research design allowing flexibility in use across different assays and studies