Learn More
Biotang Inc T4 DNA Ligase Recombinant, Cloning of restriction fragments. Joining linkers and adapters to blunt-ended DNA.
Catalog No. 50151077 Shop All BIOTANG Inc Products
Encompass_Preferred
T4 DNA Ligase Recombinant, Cloning of restriction fragments. Joining linkers and adapters to blunt-ended DNA.
Catalog No. 50-151-077 Supplier Biotang Inc Supplier No. BTENZ286
T4 DNA Ligase Recombinant, Cloning of restriction fragments. Joining linkers and adapters to blunt-ended DNA.