Learn More
Biotang Inc Thymosin b4, a 43 amino acid peptide which is regarded as the main intracellular G-actin sequestering peptide. molecular weight of 4963.55 Da, > 98%
Supplier:  Biotang Inc BTHOR275
Encompass_Preferred
Thymosin b4, a 43 amino acid peptide which is regarded as the main intracellular G-actin sequestering peptide. molecular weight of 4963.55 Da, > 98%
Catalog No. 50-151-083