Learn More
Biotang Inc Urate Oxidase Recombinant, Uricase,The specific activity was found to be 10U/mg.ontaining 302 amino acids, having a molecular formula of C1523H2383N417O462S7 and a molecular mass of 34,247 Dalton.
Supplier:  Biotang Inc BTENZ312
Encompass_Preferred
Urate Oxidase Recombinant, Uricase,The specific activity was found to be 10U/mg.ontaining 302 amino acids, having a molecular formula of C1523H2383N417O462S7 and a molecular mass of 34,247 Dalton.
Catalog No. 50-151-087