Learn More
ATCC REC-1; Mantle Cell Lymphoma; Human (Homo sapiens)
Catalog No. 502384865
Encompass
REC-1 is a lymphoblast cell line isolated in 2003 from the lymph node of a 57-year-old male with mantle cell lymphoma. The human B-cell line Rec-1 was established from a t(11;14)(q13;q32) bearing lymphoma. It can be used for immunology research.
Catalog No. 50-238-4865 Supplier ATCC Supplier No. CRL3004
REC-1 is a lymphoblast cell line isolated in 2003 from the lymph node of a 57-year-old male with mantle cell lymphoma. The human B-cell line Rec-1 was established from a t(11;14)(q13;q32) bearing lymphoma. It can be used for immunology research.