Learn More
Aviva Systems Biology Corp REC8 Antibody - N-terminal region (ARP48037_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48037_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against REC8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human REC8. Alternate names: HR21spB, MGC950, REC8L1, Rec8pApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-53534