Learn More
ABclonal Technology REC8 Rabbit pAb
SDP
Catalog No. 5000106148
Encompass_Preferred
Predicted to enable chromatin binding activity. Acts upstream of or within several processes, including gamete generation, seminiferous tubule development, and synaptonemal complex assembly. Located in kinetochore, lateral element, and male germ cell nucleus. Part of nuclear meiotic cohesin complex. Is expressed in several structures, including alimentary system, central nervous system, genitourinary system, respiratory system, and sensory organ. Orthologous to human REC8 (REC8 meiotic recombination protein).
Catalog No. 50-001-06148 Supplier ABclonal Technology Supplier No. A15002500UL
Predicted to enable chromatin binding activity. Acts upstream of or within several processes, including gamete generation, seminiferous tubule development, and synaptonemal complex assembly. Located in kinetochore, lateral element, and male germ cell nucleus. Part of nuclear meiotic cohesin complex. Is expressed in several structures, including alimentary system, central nervous system, genitourinary system, respiratory system, and sensory organ. Orthologous to human REC8 (REC8 meiotic recombination protein).