Learn More
Perkin Elmer US LLC Recalibration Service for Flowmeter Plus
Catalog No. 502105126
Encompass_Preferred
The PerkinElmer Flowmeter Plus is a valuable tool for troubleshooting detector problems. Measuring gas volumetrically eliminates there is no need to select gas type. Highly accurate, identifying leaks by differences in thermal conductivity, it offers a minimum detection limit of 0.0005 mL/min hydrogen and helium. Recalibration is hassle-free and involves only replacing the cartridge.
Catalog No. 50-210-5126 Supplier Perkin Elmer US LLC Supplier No. N9307088
The PerkinElmer Flowmeter Plus is a valuable tool for troubleshooting detector problems. Measuring gas volumetrically eliminates there is no need to select gas type. Highly accurate, identifying leaks by differences in thermal conductivity, it offers a minimum detection limit of 0.0005 mL/min hydrogen and helium. Recalibration is hassle-free and involves only replacing the cartridge.