Learn More
Recathco RECATHCO 32G IT CATHETER
Catalog No. 50196374
Encompass_Preferred
32G Intrathecal Cth Sys : contains one (7.0) 32g PU catheter ,one (8.0) PTFE coated stylet and one (27G) blunt luer stub adaptor. Dead volume 5.8 microliters,sterilized with Ethylene Oxide and ready for immediate use.
Catalog No. 50-196-374 Supplier Recathco Supplier No. 0046EO
32G Intrathecal Cth Sys : contains one (7.0) 32g PU catheter ,one (8.0) PTFE coated stylet and one (27G) blunt luer stub adaptor. Dead volume 5.8 microliters,sterilized with Ethylene Oxide and ready for immediate use.