Learn More
Recathco RECATHCO 32G OSMOTIC PUMP
Catalog No. 50196376
Encompass_Preferred
32G Intrathecal catheter that adapts to osmotic mini pump. Contains one (10.50) 32g PU catheter and one (11.0) PTFE coated stylet. Dead vol. 58.6 microliters, EtO sterilized and ready for immediate use.
Catalog No. 50-196-376 Supplier Recathco Supplier No. 0075EO
32G Intrathecal catheter that adapts to osmotic mini pump. Contains one (10.50) 32g PU catheter and one (11.0) PTFE coated stylet. Dead vol. 58.6 microliters, EtO sterilized and ready for immediate use.