Learn More
Recathco RECATHCO MOUSE CATHETER
Catalog No. 50196385
Encompass
28G Mouse vascular catheter: contains (1) 2.5 28G Mouse Vascular Catheter with (1) 4.0 .003 PTFE coated stylet. EtO sterilized and ready for use.
Catalog No. 50-196-385 Supplier Recathco Supplier No. 0189EO
28G Mouse vascular catheter: contains (1) 2.5 28G Mouse Vascular Catheter with (1) 4.0 .003 PTFE coated stylet. EtO sterilized and ready for use.