Learn More
Recathco RECATHCO MOUSE FAC
Catalog No. 50196383
Encompass_Preferred
32G Mouse Femoral Artery Catheter: contains (1) 4.5, 32G Mouse Femoral Artery Catheter with 0.003 PTFE coated stylet. EtO sterilized and ready for use
Catalog No. 50-196-383 Supplier Recathco Supplier No. 0202EO
32G Mouse Femoral Artery Catheter: contains (1) 4.5, 32G Mouse Femoral Artery Catheter with 0.003 PTFE coated stylet. EtO sterilized and ready for use