Learn More
Recathco RECATHCO MOUSE TVC
Catalog No. 50196387
Encompass_Preferred
29G Mouse Tail Vein Catheter: contains (1) Tail Vein Catheter with 29G ss needle cannula and fixed hub. EtO sterilized and ready for use
Catalog No. 50-196-387 Supplier Recathco Supplier No. 0099EO
29G Mouse Tail Vein Catheter: contains (1) Tail Vein Catheter with 29G ss needle cannula and fixed hub. EtO sterilized and ready for use