Learn More
Recathco RECATHCO Y CONNECTOR
Catalog No. 50196389
Encompass_Preferred
Rat Y Connector Catheter: contains (1) 2.0, 3F double lumen catheter adapted to 4F single lumen catheter. EtO sterilized and ready for use.
Catalog No. 50-196-389 Supplier Recathco Supplier No. 0225EO
Rat Y Connector Catheter: contains (1) 2.0, 3F double lumen catheter adapted to 4F single lumen catheter. EtO sterilized and ready for use.