Learn More
Ace Glass, Inc. Receiver, Kuderna-Danish, 10mL, 19/22, hooks, 0.1mL graduations first 1mL, 0.2mL thereafter
SDP
Catalog No. 50973384
Encompass_Preferred
10mL Kuderna-Danish receiver with a 19/22 joint and hooks. Receiver has 0.1mL graduations for the first mL, then 0.2mL graduations therafter. For concentration of materials in volatile solvents. Combines rapidity, simplicity, and quantitative efficiency, including minimization of entrainment escape and transfer losses.
Catalog No. 50-973-384 Supplier Ace Glass, Inc. Supplier No. 670837
10mL Kuderna-Danish receiver with a 19/22 joint and hooks. Receiver has 0.1mL graduations for the first mL, then 0.2mL graduations therafter. For concentration of materials in volatile solvents. Combines rapidity, simplicity, and quantitative efficiency, including minimization of entrainment escape and transfer losses.