Learn More
Ace Glass, Inc. Receiver, Kuderna-Danish, 10mL, 19/22, hooks, 0.1mL graduations first 1mL, 1mL thereafter
SDP
Catalog No. 50973385
Encompass_Preferred
10mL Kuderna-Danish receiver with a 19/22 joint and hooks. Receiver has 0.1mL graduations for the first mL, then 1mL graduations therafter. For concentration of materials in volatile solvents. Combines rapidity, simplicity, and quantitative efficiency, including minimization of entrainment escape and transfer losses.
Catalog No. 50-973-385 Supplier Ace Glass, Inc. Supplier No. 670838
10mL Kuderna-Danish receiver with a 19/22 joint and hooks. Receiver has 0.1mL graduations for the first mL, then 1mL graduations therafter. For concentration of materials in volatile solvents. Combines rapidity, simplicity, and quantitative efficiency, including minimization of entrainment escape and transfer losses.