Learn More
Ace Glass, Inc. Receiver, Kuderna-Danish, 10mL, 19/22, no hooks, 0.1mL graduations first 1mL, 0.2mL thereafter
SDP
Catalog No. 50973382
Encompass_Preferred
10mL Kuderna-Danish receiver with a 19/22 joint, and without hooks. Receiver has 0.1mL graduations for the first mL, then 0.2mL graduations therafter. For concentration of materials in volatile solvents. Combines rapidity, simplicity, and quantitative efficiency, including minimization of entrainment escape and transfer losses.
Catalog No. 50-973-382 Supplier Ace Glass, Inc. Supplier No. 670827
10mL Kuderna-Danish receiver with a 19/22 joint, and without hooks. Receiver has 0.1mL graduations for the first mL, then 0.2mL graduations therafter. For concentration of materials in volatile solvents. Combines rapidity, simplicity, and quantitative efficiency, including minimization of entrainment escape and transfer losses.