Learn More
Ace Glass, Inc. Receiver, Kuderna-Danish, 15mL, 19/22, hooks, ungraduated
SDP
Catalog No. 50973379
Encompass_Preferred
15mL Kuderna-Danish ungraduated receiver with hooks and a 19/22 joint. For concentration of materials in volatile solvents. Combines rapidity, simplicity, and quantitative efficiency, including minimization of entrainment escape and transfer losses.
Catalog No. 50-973-379 Supplier Ace Glass, Inc. Supplier No. 670815
15mL Kuderna-Danish ungraduated receiver with hooks and a 19/22 joint. For concentration of materials in volatile solvents. Combines rapidity, simplicity, and quantitative efficiency, including minimization of entrainment escape and transfer losses.