Learn More
Ace Glass, Inc. Receiver, Kuderna-Danish, 5mL, 19/22, hooks, 0.1mL graduations first 1mL, 0.2 thereafter
SDP
Catalog No. 50973383
Encompass_Preferred
5mL Kuderna-Danish receiver with a 19/22 joint and hooks. Receiver has 0.1mL graduations for the first mL, then 0.2mL graduations therafter. For concentration of materials in volatile solvents. Combines rapidity, simplicity, and quantitative efficiency, including minimization of entrainment escape and transfer losses.
Catalog No. 50-973-383 Supplier Ace Glass, Inc. Supplier No. 670835
5mL Kuderna-Danish receiver with a 19/22 joint and hooks. Receiver has 0.1mL graduations for the first mL, then 0.2mL graduations therafter. For concentration of materials in volatile solvents. Combines rapidity, simplicity, and quantitative efficiency, including minimization of entrainment escape and transfer losses.