Learn More
Ace Glass, Inc. Receiver, Kuderna-Danish, 5mL, no hooks, 0.1mL graduations first1mL, 0.2 thereafter
SDP
Catalog No. 50973381
Encompass_Preferred
5mL Kuderna-Danish receiver with a 19/22 joint, and without hooks. Receiver has 0.1mL graduations for the first mL, then 0.2mL graduations therafter. For concentration of materials in volatile solvents. Combines rapidity, simplicity, and quantitative efficiency, including minimization of entrainment escape and transfer losses.
Catalog No. 50-973-381 Supplier Ace Glass, Inc. Supplier No. 670826
5mL Kuderna-Danish receiver with a 19/22 joint, and without hooks. Receiver has 0.1mL graduations for the first mL, then 0.2mL graduations therafter. For concentration of materials in volatile solvents. Combines rapidity, simplicity, and quantitative efficiency, including minimization of entrainment escape and transfer losses.