Learn More
Ace Glass, Inc. Receiver, moisture test, 10mL, 29/42
SDP
Catalog No. 50979433
Encompass_Preferred
10mL moisture test receiver with 29/42 standard taper joint. Suitable for ASTM D 95 and E 123. Subdivided 0.1mL to 2.0mL, and 0.2mL above 2.0mL.
Catalog No. 50-979-433 Supplier Ace Glass, Inc. Supplier No. 772020
10mL moisture test receiver with 29/42 standard taper joint. Suitable for ASTM D 95 and E 123. Subdivided 0.1mL to 2.0mL, and 0.2mL above 2.0mL.