Learn More
Ace Glass, Inc. Receiver, moisture test, 10mL, 35/20
SDP
Catalog No. 50975335
Encompass_Preferred
10mL moisture test receiver with 35/20 spherical joint. Suitable for ASTM D 95 and E 123. Subdivided 0.1mL to 2.0mL, and 0.2mL above 2.0mL.
Catalog No. 50-975-335 Supplier Ace Glass, Inc. Supplier No. 772008
10mL moisture test receiver with 35/20 spherical joint. Suitable for ASTM D 95 and E 123. Subdivided 0.1mL to 2.0mL, and 0.2mL above 2.0mL.