Learn More
Ace Glass, Inc. Receiver, moisture test, 25mL, 24/40
SDP
Catalog No. 50975334
Encompass_Preferred
25mL moisture test receiver with 24/40 standard taper joint. Suitable for ASTM D 95 and E 123. Subdivided 0.1mL to 2.0mL, and 0.2mL above 2.0mL.
Catalog No. 50-975-334 Supplier Ace Glass, Inc. Supplier No. 772004
25mL moisture test receiver with 24/40 standard taper joint. Suitable for ASTM D 95 and E 123. Subdivided 0.1mL to 2.0mL, and 0.2mL above 2.0mL.