Learn More
BUCHI Receiving Flask, 10 L
SDP
Catalog No. 05405219 Shop All BUCHI Products
Change view
Click to view available options
Volume (Metric):
10 L
Catalog No. Volume (Metric)
05-405-219 10 L
1 options
Catalog No. 05-405-219 Supplier BUCHI Supplier No. 037569
Add to Cart
Add to Cart
Receiving flask for use with industrial rotary evaporators, features a P+G safety coating. Ideal for collecting condensed solvents.
  • Exclusive use of DURAN™ borosilicate glass 3.3
  • Maximum safety guaranteed by safety coated glassware (P+G)
  • High chemical resistance against acids, alkalis, and organic substances
Specifications
Material Borosilicate Glass (3.3)
Type Receiving Flask
For Use With (Application) Collecting Condensed Solvents
For Use With (Equipment) R-220 Pro, R-220 Essential, R-220
Volume (Metric) 10 L