Learn More
BUCHI Receiving Vessel, 420 mL
SDP
Catalog No. 05405655 Shop All BUCHI Products
Change view
Click to view available options
Capacity (Metric):
420 mL
Catalog No. Capacity (Metric)
05-405-655 420 mL
1 options
Catalog No. 05-405-655 Supplier BUCHI Supplier No. 043390
Add to Cart
Add to Cart
Receiving Vessel for use with K-350, K-355, K-360, and K-375.
  • Capacity: 420 mL
Specifications
Type Receiving Vessel
Capacity (Metric) 420 mL
For Use With (Equipment) K-350, K-355, K-360, and K-375