Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO Receptacle -100 ml GL reactor

Catalog No. NC1434245 Shop All METTLER TOLEDO Products
Encompass

Receptacle for 100 ml glass reactor

Catalog No. NC1434245 Supplier METTLER TOLEDO Supplier No. 30086802
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Receptacle for 100 ml glass reactor

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.