Learn More
Biomatik Corporation Receptor Activator Of Nuclear Factor Kappa B Ligand (Rankl) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29312
Encompass_Preferred
Receptor Activator Of Nuclear Factor Kappa B Ligand (Rankl) Monoclonal Antibody is a Rat (Rattus norvegicus) Monoclonal Antibody to Receptor Activator Of Nuclear Factor Kappa B Ligand (RANkL). The Receptor Activator Of Nuclear Factor Kappa B Ligand (Rankl) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Rat (Rattus norvegicus). Immunogen: Glu106~Met240. Applications: WB, IHC, ICC, IP. Target Synonyms: Receptor Activator Of Nuclear Factor Kappa B Ligand (RANkL). Accession Number: Q9ESE2. Gene ID: 117516. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5923