Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Receptor tyrosine-protein kinase erbB-3 (ErbB3/HER3) | >95.0% | 10 UG
SDP

Catalog No. 5000291388
Encompass_Preferred

Recombinant rat HER3 protein (residues S20-H641) expressed in HEK293 cells with a C-terminal His tag. The protein is glycosylated and has an apparent molecular weight of approximately 85-115 kDa. Supplied at high purity (>95% by reducing SDS-PAGE) for biochemical assays, binding studies, and antibody development.

  • Expression system: HEK293 cells
  • Tag: C-terminal His tag
  • Sequence region: residues S20-H641
  • Molecular weight: ~85-115 kDa (glycosylated)
  • Purity: >95% by reducing SDS-PAGE
  • Applications: biochemical assays, binding studies, antibody development

Catalog No. 50-002-91388 Supplier Medchemexpress LLC Supplier No. HYP7489110UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat HER3 protein (residues S20-H641) expressed in HEK293 cells with a C-terminal His tag. The protein is glycosylated and has an apparent molecular weight of approximately 85-115 kDa. Supplied at high purity (>95% by reducing SDS-PAGE) for biochemical assays, binding studies, and antibody development.

  • Expression system: HEK293 cells
  • Tag: C-terminal His tag
  • Sequence region: residues S20-H641
  • Molecular weight: ~85-115 kDa (glycosylated)
  • Purity: >95% by reducing SDS-PAGE
  • Applications: biochemical assays, binding studies, antibody development

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.