Learn More
Grainger Westward Recessed Grease Extension Adapter, Flush-Style, Zerk Connection, 3,000 psi, 1 ea
Catalog No. 502771900
Encompass_Preferred
This recessed lube greasing adapter is a flush-style coupler designed to provide a secure, leak-tight seal when connecting to zerk fittings. It is engineered for use with hand-operated grease guns to facilitate effective lubrication in recessed areas.
  • Flush-style coupler design
  • Compatible with zerk fittings
  • Rated for a maximum operating pressure of 3,000 psi
  • Includes an extension nozzle and locking sleeve
Catalog No. 50-277-1900 Supplier Grainger Supplier No. 13X055
This recessed lube greasing adapter is a flush-style coupler designed to provide a secure, leak-tight seal when connecting to zerk fittings. It is engineered for use with hand-operated grease guns to facilitate effective lubrication in recessed areas.
  • Flush-style coupler design
  • Compatible with zerk fittings
  • Rated for a maximum operating pressure of 3,000 psi
  • Includes an extension nozzle and locking sleeve