Learn More
Chemglass Life Sciences Recessed Platform (Fig. 5)
SDP
Catalog No. 5000083116
Encompass
NOTE: VORTEX-GENIE 1 MAY ONLY BE USED WITH POP-OFF CUP AND 3" PLATFORM.
Catalog No. 50-000-83116 Supplier Chemglass Life Sciences Supplier No. CLS4891005
NOTE: VORTEX-GENIE 1 MAY ONLY BE USED WITH POP-OFF CUP AND 3" PLATFORM.