Learn More
Fisherbrand™ Recessed Safety Station
Catalog No. 15050312 Shop All Fisherbrand Products
Change view
Click to view available options
Mounting Type:
Ceiling
Wall
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Type
15-050-312 Wall
15-050-311 Ceiling
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 15-050-312
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
Recessed barrier-free eye/face wash and shower safety station with ceiling or wall mounted exposed shower head and drain pan.
  • Recessed barrier-free eye/face wash and shower safety station with ceiling or wall mounted exposed shower head and drain pan.
  • 10 in. diameter stainless steel shower head with 20 GPM flow control.
  • 1 in. IPS brass stay-open shower ball valve with stainless steel 'panic bar' actuator.
Specifications
Product Type Safety Station
Material Brass
Mounting Type Wall
WARNING: Cancer - www.P65Warnings.ca.gov