Learn More
Gibco™ Recharge™ Supplement, Without Glucose and L-glutamine

Description
- A chemically defined (CD) supplement designed to increase protein yields, while maintaining acceptable viability and cell growth
- The supplement has been developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors
- In order to increase versatility of the formulation, glucose and L-glutamine are not included in the final formulations
- Therefore, these two components should be supplemented by the user and monitored as appropriate
- The Recharge™ Supplement was developed for use in CHO mammalian cell culture systems and can be used either as a media supplement or a culture system feed
- However, user data have shown this CD supplement has been beneficial for non-CHO mammalian systems and microbial systems
- Additionally, the Recharge™ Supplement has been successfully used as a replacement for yeast extract
Specifications
Specifications
| Content And Storage | Store in cool, dry conditions in well-sealed containers, 2°C to 8°C |
| Form | Powder |
| Product Type | CHO Media Supplement |
| For Use With (Application) | Mammalian Cell Culture |
| Quantity | 1 kg |
| Source | Animal origin-free |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.