Learn More
Med Vet International Rechargeable Batteries - Welch Allyn Compatible - 2.5v 600mAh NiCad, J1663, Each
Catalog No. 5000583250
Encompass_Preferred
JorVet offers a range of quality rechargeable cells for use with Welch Allyn handles. Guaranteed for 2 years with an average working life of 5 years. Battery Type: 2.5v 600mAh NiCad WA Code: 7200 Label: Red
Catalog No. 50-005-83250 Supplier Med Vet International Supplier No. JOR1663
JorVet offers a range of quality rechargeable cells for use with Welch Allyn handles. Guaranteed for 2 years with an average working life of 5 years. Battery Type: 2.5v 600mAh NiCad WA Code: 7200 Label: Red