Learn More
Med Vet International Rechargeable Batteries - Welch Allyn Compatible - 2.5v 800mAh NiCad, J1663C, Each
Catalog No. 5000583251
Encompass_Preferred
JorVet offers a range of quality rechargeable cells for use with Welch Allyn handles. Guaranteed for 2 years with an average working life of 5 years. Battery Type: 2.5v 600mAh NiCad WA Code: 7260 Label: Blue
Catalog No. 50-005-83251 Supplier Med Vet International Supplier No. JOR1663C
JorVet offers a range of quality rechargeable cells for use with Welch Allyn handles. Guaranteed for 2 years with an average working life of 5 years. Battery Type: 2.5v 600mAh NiCad WA Code: 7260 Label: Blue