Learn More
Med Vet International Rechargeable Batteries - Welch Allyn Compatible - 3.5v 800mAh NiCad, J1663B, Each
Catalog No. 5000583558
Encompass_Preferred
JorVet offers a range of quality rechargeable cells for use with Welch Allyn handles. Guaranteed for 2 years with an average working life of 5 years. Battery Type: 3.5v 800mAh NiCad WA Code: 7230 Label: Orange
Catalog No. 50-005-83558 Supplier Med Vet International Supplier No. JOR1663B
JorVet offers a range of quality rechargeable cells for use with Welch Allyn handles. Guaranteed for 2 years with an average working life of 5 years. Battery Type: 3.5v 800mAh NiCad WA Code: 7230 Label: Orange