Learn More
Lovibond™ Rechargeable Battery Pack
Catalog No. 09051518 Shop All Tintometer Group Products
Change view
Click to view available options
For Use With (Equipment):
Turbidimeter
Catalog No. For Use With (Equipment)
09-051-518 Turbidimeter
1 options
Catalog No. 09-051-518 Supplier Lovibond™ Supplier No. 19820011
Add to Cart
Add to Cart
This battery pack, Ni-MH can be used with the Lovibond TB350 instead of standard AA batteries for convenience and increased run time.
  • This battery pack is suitable for use in TB350 instruments with serial number 23041000 or higher. For earlier serial numbers, please contact Lovibond Support
  • Battery pack charges while the unit is plugged in via USB-C. An optional power supply is available separately
Specifications
Type Batteries
For Use With (Equipment) Turbidimeter