Learn More
ACCU-SCOPE Rechargeable Binocular Microscope with 3 Objectives
SDP
Catalog No. 01189313 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Magnification Power:
4X, 10X, Semi-Plan 40XR
Catalog No. Magnification Power
01-189-313 4X, 10X, Semi-Plan 40XR
1 options
Catalog No. 01-189-313 Supplier ACCU-SCOPE Supplier No. EXC1203RC
Add to Cart
Add to Cart
Offers reliable, high-quality viewing for lab and educational use with clear images and durable construction for daily use.
  • Binocular viewing head inclined 30° rotatable 360°
  • WF10X18.5 mm eyepiece paired one wpointer diopter adjustment
  • Reversed quadruple nosepiece
  • DIN 4X 10X and SP40XR objectives
  • Rechargeable LED base
  • Rackless mechanical stage w low position coaxial controls
  • Abbe condenser NA 1.25 with iris diaphragm and filter holder
  • Coaxial coarse and fine focusing controls
  • Variable LED illuminator UL listed
  • Dust cover power cord and manual
Specifications
Eyepiece WF 10X/18.5 mm
Head Style Binocular, 30° Inclined, 360° Rotating
Lighting Type LED
Magnification Power 4X, 10X, Semi-Plan 40XR
Model ACCU-SCOPE
Objective Types DIN Achromatic
Product Type Binocular LED Microscope
Voltage 110/240 V