Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Rechargeable Li-Ion Battery Pack for M210 Handheld Label Maker

Catalog No. 19000746 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-000-746 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-000-746 Supplier Brady™ Supplier No. M210BATTERY
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Get longer battery life and save money on AA batteries with Brady™ M210 Rechargeable Li-Ion Battery Pack.

  • Get longer battery life and save money on AA batteries with this M210 Rechargeable Li-Ion Battery Pack.
  • Package includes spare printer battery (label printer and charger sold separately).
  • Long battery life; prints at least 2,000 2 in. (50.8 mm) long labels per charge (battery charges outside of printer using AC adaptor).
  • Use AC adapter/charger to charge battery (not included).
  • Enables cordless printer operation to print labels where (and when) you need them.

Specifications

Product Type Printer
Color Gray
Display Type None
For Use With (Equipment) Brady Labels, Brady M210-LAB Label Printer, BMP21 LAB Label Printer, BMP21-PLUS Label Printer
Material Plastic
Width (English) 11.5 in.
Sterility Non-sterile
Power Supply Details Lithium-Ion Battery (Must Ship Ground)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.