Learn More
Brady Rechargeable NiMH Battery for M410 Printer Label Printer
Catalog No. 17467582 Shop All Brady Corporation Products
Change view
Click to view available options
For Use With (Equipment):
BMP41 Label Printer, Brady M410 Label Printer, BMP61 Labeling System
Catalog No. For Use With (Equipment)
17-467-582 BMP41 Label Printer, Brady M410 Label Printer, BMP61 Labeling System
1 options
Catalog No. 17-467-582 Supplier Brady Supplier No. M410BATT
  • $158.00
    $152.65 / Each of 1
    Save $5.35
    3% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Get longer battery life and save money on AA batteries with this rechargeable NiMH battery for the M410 Label Printer. Printer can be run while battery is charging.
  • Package includes spare Printer Battery only (label printer and charger sold separately).
  • Long battery life - prints up to 1000 1 in. x 2 in. labels per battery charge.
  • Use AC adapter/charger unit (not included) to charge battery.
  • Enables cordless printer operation so you can print labels where you need them, when you need them.
Specifications
Product Type Battery/Adapter Pack
For Use With (Equipment) BMP41 Label Printer, Brady M410 Label Printer, BMP61 Labeling System