Learn More
Hillrom Rechargeable Power Handle
Catalog No. 22355141
Change view
Click to view available options
For Use With (Equipment):
IEC Plug Type-A
Universal Desk Charger
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
22-355-141 Universal Desk Charger
22-355-148 IEC Plug Type-A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-355-141 Supplier Hillrom Supplier No. WEL 71670
Add to Cart
Add to Cart
This item is not returnable. View return policy
Nickel-Cadmium Rechargeable Handles - A choice of convenient rechargeable power sources that fit all 3.5 V instrument heads.
  • Rechargeable nickel-cadmium battery included
  • Smooth, nonabrasive knurled finish ensures a no-slip grip
  • Chrome-plated brass construction for durability and reliable long-life
  • Recharges in our desk charger, stand consoles, and well-chargers
Specifications
Voltage 3.5 V
Disposable Reusable
Product Type Rechargeable Otoscope Handle
For Use With (Application) Physical Exams
For Use With (Equipment) Universal Desk Charger
Includes Nickel-Cadmium Battery