Learn More
Stoelting™ Rechargeable Surgical Clippers
SDP
Catalog No. 10000789 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Cordless, rechargable
Catalog No. Model
10-000-789 Cordless, rechargable
1 options
Catalog No. 10-000-789 Supplier Stoelting™ Supplier No. 51465
Add to Cart
Add to Cart
Stoelting™ Rechargeable Surgical Clippers is a powerful, detachable blade clipper designed for quick and easy results for all-day heavy use.
  • Quiet operation-will not frighten sensitive animals
  • High-speed motor-3800 strokes per minute
  • Runs continuously for 60 to 75 minutes with fully charged battery pack
  • Cordless operation provides easy mobility around animals
  • Hair won't be blown into wounds or incisions-operates cool without fans.
  • Virtually maintenance-free with no oiling or greasing of internal parts needed
  • On/Off switch with lock for safe operation
  • Heavy duty storage case holds and protects all components
Specifications
For Use With (Application) Neuroscience
Size #40 blade
Model Cordless, rechargable