Learn More
ACCU-SCOPE Rechargeable Trinocular Microscope with Achromat Objectives
SDP
Catalog No. 01189344 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Magnification Power:
4X, 10X, SP40XR, 100XR
Catalog No. Magnification Power
01-189-344 4X, 10X, SP40XR, 100XR
1 options
Catalog No. 01-189-344 Supplier ACCU-SCOPE Supplier No. EXC123PLRC
Add to Cart
Add to Cart
Offers reliable viewing for labs and classrooms with durable construction and clear image quality.
  • Trinocular viewing head and photo tube inclined 30° rotatable 360°
  • WF10X18.5 mm eyepiece paired one wpointer diopter adjustment
  • Reversed quadruple nosepiece
  • Plan achromat objectives 4X 10X SP40X R and 100X R oil objectives
  • Rackless mechanical stage w low position coaxial controls
  • Abbe condenser NA 1.25 with iris diaphragm and filter holder
  • Coaxial coarse and fine focusing controls
  • Variable LED illuminator UL listed
  • Dust cover power cord and manual
Specifications
Eyepiece WF 10X/18.5 mm
Head Style Trinocular, 30° Inclined, 360° Rotating
Lighting Type LED
Magnification Power 4X, 10X, SP40XR, 100XR
Model ACCU-SCOPE
Objective Types Plan Achromatic
Product Type Trinocular LED Microscope
Voltage 110/240 V