Learn More
TARGETMOL CHEMICALS INC RECILISIB 10MG
SDP
Catalog No. 502577390
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Recilisib (ON 01210) is a radioprotectantcan activate AKT PI3K activities in cells. purity: 99%
Catalog No. 50-257-7390 Supplier TARGETMOL CHEMICALS INC Supplier No. T1386210MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Recilisib (ON 01210) is a radioprotectantcan activate AKT PI3K activities in cells. purity: 99%