Learn More
Medchemexpress LLC Recilisib | 334969-03-8 | MFCD07366276 | 99.9% | 336.79 g/mol | C16H13ClO4S | 50 MG
SDP
Catalog No. 5000248210
Encompass_Preferred
Recilisib (ON 01210) is a small-molecule radioprotectant reported to activate AKT and PI3K signaling in cells. It is supplied as a high-purity research reagent with recommended solvent guidance and multiple packaging formats for in vitro and in vivo studies.
  • High purity suitable for research use.
  • Activates AKT and PI3K signaling in cellular assays.
  • Soluble in DMSO; protocols available for in vitro and in vivo dosing.
  • Offered as solids, solutions, and sample formats.
  • Multiple package sizes to support screening and scale-up.
Catalog No. 50-002-48210 Supplier Medchemexpress LLC Supplier No. HY10162550MG
Recilisib (ON 01210) is a small-molecule radioprotectant reported to activate AKT and PI3K signaling in cells. It is supplied as a high-purity research reagent with recommended solvent guidance and multiple packaging formats for in vitro and in vivo studies.
  • High purity suitable for research use.
  • Activates AKT and PI3K signaling in cellular assays.
  • Soluble in DMSO; protocols available for in vitro and in vivo dosing.
  • Offered as solids, solutions, and sample formats.
  • Multiple package sizes to support screening and scale-up.