Learn More
Medchemexpress LLC Recilisib (ON 01210) | 334969-03-8 | MFCD07366276 | 99.9% | 336.79 g/mol | C16H13ClO4S | 25 MG
SDP
Catalog No. 5000248217
Encompass_Preferred
Recilisib is a small-molecule radioprotectant that activates PI3K and AKT signaling to protect hematopoietic progenitor cells from radiation-induced damage. It preserves colony-forming ability of bone marrow cells in vitro and enhances recovery of myeloid progenitors in vivo. Chemical properties include formula C16H13ClO4S and molecular weight 336.79 g/mol.
  • Acts as a radioprotectant by activating PI3K and AKT signaling in cells.
  • Protects bone marrow colony-forming units from radiation-induced depletion.
  • Enhances recovery and differentiation of primitive myeloid progenitors in vivo.
  • Solid, white to off-white form with DMSO solubility and established in vivo formulations.
  • High purity (≈99.9%) suitable for research use.
Catalog No. 50-002-48217 Supplier Medchemexpress LLC Supplier No. HY10162525MG
Recilisib is a small-molecule radioprotectant that activates PI3K and AKT signaling to protect hematopoietic progenitor cells from radiation-induced damage. It preserves colony-forming ability of bone marrow cells in vitro and enhances recovery of myeloid progenitors in vivo. Chemical properties include formula C16H13ClO4S and molecular weight 336.79 g/mol.
  • Acts as a radioprotectant by activating PI3K and AKT signaling in cells.
  • Protects bone marrow colony-forming units from radiation-induced depletion.
  • Enhances recovery and differentiation of primitive myeloid progenitors in vivo.
  • Solid, white to off-white form with DMSO solubility and established in vivo formulations.
  • High purity (≈99.9%) suitable for research use.