Learn More
Across International Ai -15°C 3L Compact Recirculating Chiller with Centrifugal Pump
SDP
Supplier:  Across International C1532LUL.110
Encompass_Preferred
Ai C15 series recirculating chillers are the true thermostatic control laboratory chiller, this closed laboratory chiller is ideal for applications such as cooling rotary evaporators, analytical instrumentation, laser etching, ICP, vacuum systems, plasma etching and jacketed reactors. This system can also be used as a replacement to tap water cooling in laboratory applications.
Catalog No. NC1971795