Learn More
Being Scientific Recirculating Chiller 0.5 cuft (15.5L) 120V, 1800W Cooling capacity
Catalog No. 5000217588
Encompass_Preferred
The BEING BRC Series cooling/heating recirculating chillers provide continuously reliable and stable laboratory temperature control for a wide range of applications. The recirculating chillers have a bright, easy-to-understand LCD intelligent controller, a corrosion-resistant stainless steel chamber, and many more features. Available in 3 L, 10 L, and 20 L tank capacity with a temperature range from -20oC to 20oC.
Catalog No. 50-002-17588 Supplier Being Scientific Supplier No. BRC15110U
The BEING BRC Series cooling/heating recirculating chillers provide continuously reliable and stable laboratory temperature control for a wide range of applications. The recirculating chillers have a bright, easy-to-understand LCD intelligent controller, a corrosion-resistant stainless steel chamber, and many more features. Available in 3 L, 10 L, and 20 L tank capacity with a temperature range from -20oC to 20oC.