Learn More
Being Scientific Recirculating Chiller 1.1 cuft 30L) 240V, 2800W Cooling capacity
Catalog No. 5000217537
Encompass_Preferred
The BEING BRC Series cooling/heating recirculating chillers provide continuously reliable and stable laboratory temperature control for a wide range of applications. The recirculating chillers have a bright, easy-to-understand LCD intelligent controller, a corrosion-resistant stainless steel chamber, and many more features. Available in 3 L, 10 L, and 20 L tank capacity with a temperature range from -20oC to 20oC.
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 50-002-17537 Supplier Being Scientific Supplier No. BRC15120C
The BEING BRC Series cooling/heating recirculating chillers provide continuously reliable and stable laboratory temperature control for a wide range of applications. The recirculating chillers have a bright, easy-to-understand LCD intelligent controller, a corrosion-resistant stainless steel chamber, and many more features. Available in 3 L, 10 L, and 20 L tank capacity with a temperature range from -20oC to 20oC.
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.